missing translation for 'onlineSavingsMsg'
Learn More

CD38 Mouse anti-Human, PE, Clone: HB7, BD

Product Code. p-7092545
Change view
Click to view available options
Cantidad:
50 Tests
Unit Size:
cada uno
Product Code. Cantidad unitSize
15825988 50 Tests cada uno
1 options
This item is not returnable. View return policy

Product Code. 15825988

Brand: BD Biosciences 342371

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD38, clone HB7, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the BJAB cell line.

CD38 recognizes an integral membrane glycoprotein, Mr 45 kilodaltons (kd), with a protein core of 35 kd.

TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD38
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon HB7
Concentración 12.5μg/mL
Conjugado PE
Formulación Phosphate buffered saline with BSA and 0.1% sodium azide.
Especie del huésped Mouse
Inmunógeno Human BJAB B cell line
Cantidad 50 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.