Learn More
CD38 Mouse anti-Human, PE, Clone: HB7, BD
Descripción
CD38, clone HB7, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the BJAB cell line.
CD38 recognizes an integral membrane glycoprotein, Mr 45 kilodaltons (kd), with a protein core of 35 kd.
Especificaciones
Especificaciones
| Antígeno | CD38 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | HB7 |
| Concentración | 12.5μg/mL |
| Conjugado | PE |
| Formulación | Phosphate buffered saline with BSA and 0.1% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Human BJAB B cell line |
| Cantidad | 50 Tests |
| Mostrar más |
For Research Use Only.