Learn More
CD36 Antibody (FA6-152), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Marca: Novus Biologicals NB600-1423
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD36 Monoclonal specifically detects CD36 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
Especificaciones
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Western Blot 1:100-1:2000, Flow Cytometry 2 ug/10^5 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 ug antibody/200 ug protein, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500, Functional >1 ug/ml, Immunoassay 10 ug/ml | |
| Protein G purified | |
| Purified |
| Monoclonal | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Store at 4°C. | |
| IgG1 |