missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (1A7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP3-11430AF700
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Especificaciones
| CD36 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| 1A7 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| BDPLT10, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, leukocyte differentiation antigen CD36, PAS IV, PAS-4 protein, PASIV, platelet glycoprotein 4, platelet glycoprotein IV, SCARB3, scavenger receptor class B, member 3 | |
| Human CD36 from platelets (Uniprot: P16671) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido