Learn More
CD35 Mouse anti-Human, Brilliant Violet 510, Clone: E11, BD Optibuild™
Descripción
The E11 monoclonal antibody specifically binds to CD35. CD35 is also known as Complement receptor type 1 (CR1), C3b/C4b receptor, C3BR, C4BR, Immune adherence receptor, or KN. CD35 is a type I transmembrane glycoprotein that exists in four allelic forms of 160, 190, 220 and 250 kDa. CD35 serves as a receptor for complement fragments C3b, iC3b, C3dg, C4b, iC3, and iC4. It enhances phagocytosis by neutrophils and monocytes and regulates complement activation. It is expressed on erythrocytes, granulocytes, monocytes, B cells, and some dendritic cells, T cells, and NK cells. It binds complement components C3b and C4b, mediating. This antibody cannot inhibit the phagocytic capacity of granulocytes. The CD35 antibody is useful in studies of cells that express complement receptors.
Especificaciones
Especificaciones
| Antígeno | CD35 |
| Clasificación | Monoclonal |
| Clon | E11 |
| Concentración | 0.2mg/mL |
| Conjugado | Brilliant Violet 510 |
| Formulación | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Alias de gen | CR1, Complement receptor type 1, C3b/C4b receptor, C3BR, C4BR, KN |
| Especie del huésped | Mouse |
| Inmunógeno | Human Cells of the Monocyte Lineage |
| Método de purificación | Affinity Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.