missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD34 Monoclonal Antibody (MEC14.7), PE

Código de producto. 15314111
Change view
Click to view available options
Cantidad:
300 μg
50 μg
Tamaño de la unidad:
300 microgramos
50 microgramos
Código de producto. Cantidad unitSize
15314111 300 μg 300 microgramos
15304111 50 μg 50 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15314111

Marca: Invitrogen™ MA517832

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

CD34 Monoclonal Antibody for Flow

CD34 is a highly glycosylated monomeric with a molecular weight range of 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD34
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon MEC14.7
Concentración 0.1 mg/mL
Conjugado PE
Formulación PBS with 4-5mg/mL BSA and 0.02% sodium azide
génica Cd34
N.º de referencia del gen Q64314
Alias de gen (E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1
Símbolos de los genes Cd34
Especie del huésped Rat
Inmunógeno Murine transformed endothelioma cell line t-end.
Método de purificación Protein G
Cantidad 300 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 12490
Especies diana Mouse
Contenido y almacenamiento 4°C, do not freeze
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2a
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.