missing translation for 'onlineSavingsMsg'
Learn More

CD335 (NKp46) Monoclonal Antibody (9E2), NovaFluor™ Red 685, eBioscience™, Invitrogen™

Código de producto. 17757547
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
paquete de 100
paquete de 25
Código de producto. Cantidad unitSize
17757547 100 Tests paquete de 100
17727547 25 Tests paquete de 25
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 17757547

Marca: Invitrogen H040T03R02

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

This 9E2 monoclonal antibody reacts with human CD335, which is also known as NKp46. Expressed on NK cells, CD335 is a member of the natural cytotoxicity receptor family that also includes NKp30 and NKp44. This type I transmembrane glycoprotein is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor and virus-infected (e.g., influenza) cells. CD335 also interacts with pancreatic beta cells during the development of type 1 diabetes. The 9E2 antibody has been reported to induce cytotoxicity. Applications Reported: This 9E2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human periheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Use 5 μL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 637 nm; Emission: 685 nm; Laser: 633-640 nm (Red) Laser

NKp46 (CD335, NCR1, ) is a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. NKp46 consists of two Ig-like domains assembled to leukocyte immunoglobulin-like (LIR) and killer inhibitory receptors (KIR).
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD335 (NKp46)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 9E2
Concentración 0.4 μg/Test
Conjugado NovaFluor Red 685
Formulación PBS with 0.09% sodium azide; pH 7.2
génica NCR1
N.º de referencia del gen O76036
Alias de gen 1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1
Símbolos de los genes NCR1
Especie del huésped Mouse
Método de purificación Affinity chromatography
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9437
Especies diana Human
Contenido y almacenamiento 4° C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.