missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Monoclonal Antibody (OTI1C4), TrueMAB™, OriGene
Descripción
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Immunocytochemistry, Western Blot
Especificaciones
Especificaciones
| Antígeno | CD33 |
| Aplicaciones | Immunocytochemistry, Western Blot |
| Clasificación | Monoclonal |
| Clon | OTI1C4 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| génica | CD33 |
| N.º de referencia del gen | P20138 |
| Alias de gen | CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?