Learn More
Cd31 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28083-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Cd31 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Cd31. This antibody recognizes mouse, human antigen. The Cd31 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| Cd31 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 18613 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Human |