Learn More
Epredia™ Lab Vision™ CD31/PECAM-1 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody
Tienda Epredia ProductosDescripción
Host Species: Mouse
Clone: JC/70A
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat and pig. Others not tested.
Epitope: Not determined
Immunogen: Membrane preparation of a spleen from a patient with hairy cell leukemia
Molecular Weight: 100kDa (endothelial cells) and 130kDa (platelets)
Positive Control: Placenta, Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
- Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | CD31 (Endothelial Cell Marker) Ab-1 |
| Aplicaciones | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | JC/70A |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Inmunógeno | Membrane preparation of a spleen from a patient with hairy cell leukemia |
| Cantidad | 7 mL |
| Estado normativo | IVD |
| Disciplina de investigación | Oncogenesis |
| Mostrar más |