Learn More
CD31/PECAM-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 57 publications
Marca:  Novus Biologicals NB100-2284
Código de producto. 18786833
  • ONLINE EXCLUSIVE PRICE
    698.00€
    659.40€ / 0.10 ml
    Ahorro 38.60€
    5% Descuento
Descripción
CD31/PECAM-1 Polyclonal specifically detects CD31/PECAM-1 in Human, Mouse, Rat, Porcine, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
PECAM1
Human, Mouse, Pig
Polyclonal
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Affinity Purified
IgG
Certificados
Promociones
Promociones
For Research Use Only