missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD31 Monoclonal Antibody (MBC78.2), FITC
GREENER_CHOICE

Código de producto. p-7170012
Change view
Click to view available options
Cantidad:
500 μL
2 mL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10671244 500 μL cada uno
10028632 2 mL cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10671244

Marca: Invitrogen™ MHCD3101

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.

CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD31
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon MBC78.2
Concentración 0.2mg/mL
Conjugado FITC
Descripción CD31 Monoclonal Antibody, 0.5mL, 0.2mg/mL Concentration, MBC78.2 Clone, CD31 Antigen, Human CD31 Immunogen, Mouse Host, Human Species Reactivity, FITC Conjugate, IgG1 Isotype, PBS + BSA Storage Buffer, Used for Flow Cytometry
Dilución Assay-dependent
Formulación PBS with 4mg/mL BSA and 0.1% sodium azide
génica Pecam1
N.º de referencia del gen P16284
Alias de gen C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule
Símbolos de los genes Pecam1
Especie del huésped Mouse
Inmunógeno Human CD31.
Método de purificación Purified
Cantidad 500 μL
Estado normativo ASR
Primario o secundario Primary
ID de gen (Entrez) 5175
Envase 1 Each
Incluye 0.1% Sodium azide
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.