missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody

Código de producto. 12691177 Tienda Epredia Productos
Change view
Click to view available options
Cantidad:
100 μL
500 μL
1 mL
7 mL
12 mL
Formulario:
Concentrated
Prediluted
Ready-to-Use
Tamaño de la unidad:
1 ml
100 microlitros
12 ml
500 microlitros
7 ml
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad Formulario unitSize
12691177 7 mL Prediluted 7 ml
12621187 100 μL Concentrated 100 microlitros
12631187 500 μL Concentrated 500 microlitros
12611187 1 mL Concentrated 1 ml
12601187 12 mL Ready-to-Use 12 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12691177 Proveedor Epredia™ N.º de proveedor MS361R7

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody.

CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells.

Host Species: Mouse

Clone: Ber-H2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with baboon and monkey. Others not known.

Epitope: Not determined

Immunogen: Co cells

Molecular Weight: 120 kDa (mature), 95 kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD30 Ab-1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon Ber-H2
Conjugado Unconjugated
Especie del huésped Mouse
Inmunógeno Co cells
Cantidad 7 mL
Estado normativo IVD
Disciplina de investigación Oncogenesis
Primario o secundario Primary
Especies diana Human
Formulario Prediluted
Isotype IgG2a κ
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.