Learn More
CD3 Mouse anti-Human , BV510, Clone: OKT3, BD Biosciences™
Código de producto. p-7231843
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
100 tests
25 tests
17237604 25 Tests 25 tests
17227604 100 Tests 100 tests
2 options
Código de producto. 17237604
Marca: BD Biosciences 566780
Mouse Monoclonal Antibody
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno CD3
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon OKT3
Conjugado Brilliant Violet 510
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Alias de gen CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE
Especie del huésped Mouse
Inmunógeno Sheep Erythrocyte Rosette-purified Human T Cells
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV510 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV510 were removed.
Isotype IgG2a κ