Learn More
Descripción
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human A431 whole cell, human HEK293 whole cell, human U20S whole cell, human HL-60 whole cell, human MCF-7 whole cell, human Hela whole cell, human PANC-1 whole cell, rat liver tissue, rat testicular tissue, mouse pancreas tissue, mouse testicular tissue, mouse RAW2467 whole cell. IHC: human mammary cancer, human mammary cancer, human colon cancer, human placenta tissue, mouse kidney tissue, rat brain tissue. ICC/IF: A431 cell. Flow: K562 cell, U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Especificaciones
Especificaciones
| Antígeno | CD2AP |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | 5F8 |
| Concentración | 500 μg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 4mg trehalose and 0.05mg sodium azide |
| génica | CD2AP |
| N.º de referencia del gen | F1LRS8, Q9JLQ0, Q9Y5K6 |
| Alias de gen | adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
