missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD298 Recombinant Rabbit Monoclonal Antibody (23GB4740), Invitrogen™
Descripción
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 3 subunit. This gene encodes a beta 3 subunit. A pseudogene exists for this gene, and it is located on chromosome 2.
Especificaciones
Especificaciones
| Antígeno | CD298 |
| Aplicaciones | Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB4740 |
| Concentración | 0.45 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | ATP1B3 |
| N.º de referencia del gen | P54709, P97370, Q63377 |
| Alias de gen | AA409958; AI664000; Atp1b3; ATPase Na+/K+ transporting subunit beta 3; ATPase, Na+/K+ beta 3 polypeptide; ATPase, Na+/K+ transporting, beta 3 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 3; ATPB-3; AW212096; CD298; CD298 antigen; Na K-ATPase beta-3 subunit; Na+/K+ -ATPase beta 3 subunit; NKAB3S; Sodium Potassium ATPase; Sodium/potassium-dependent ATPase subunit beta-3; sodium/potassium-transporting ATPase subunit beta-3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?