missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Monoclonal Antibody (CD28.2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Especificaciones
| Antígeno | CD28 |
|---|---|
| Clon | CD28.2 |
| Concentración | 5 μL/Test |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
Descripción
Mouse Monoclonal Antibody
CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary's Disease.Especificaciones
| CD28 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 940 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| CD28.2 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Mouse | |
| Human | |
| P10747 | |
| Cd28 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Cd28 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto