missing translation for 'onlineSavingsMsg'
Learn More

CD28 Mouse anti-Human, PE-Cy5, Clone: CD28.2, BD

Código de producto. 15848398
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15848398 100 Tests cada uno
15861339 25 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15848398 Leverandør BD Biosciences Leverandørnr. 555730

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.

TRUSTED_SUSTAINABILITY

Tekniske data

Antígeno CD28
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon CD28.2
Conjugado PE-Cyanine5
Descripción CD28 antigen; T44; Tp44; TP44
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human CD28 Transfected Cell Line
Método de purificación Affinity Purified
Cantidad 100 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.