missing translation for 'onlineSavingsMsg'
Learn More

CD271 Mouse anti-Human, PE, Clone: C40-1457, BD

Código de producto. 15810609
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15810609 25 Tests cada uno
15818858 100 Tests cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15810609 Proveedor BD Biosciences N.º de proveedor 560927

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The C40-1457 monoclonal antibody specifically binds to CD271, the nerve growth factor receptor (NGFR). CD271 is a transmembrane 75 kDa protein that has been found localized to neuronal axons, Schwann cells, and perineural cells of peripheral nerves. It is also expressed by cells in some epithelial, mesenchymal and lymphoid tissues. NGFR is the receptor for nerve growth factor (NGF), apolypeptide that is essential for normal development of the nervous system. NGF promotes survival and differentiation of sympathetic and sensory neurons during embryological development of peripheral neurons. NGF binds to two distinctive surface receptors expressed by target cells, the p140-TrkA (NTRK1) and the p75 NGFR. High affinity binding of NGF requires that both receptor molecules be expressed. NGFR has been found on human and rat lymphocytes. A subset of lymphoid cells in the spleen, lymph nodes, and follicular dentritic cells in germinal centers of reactive lymph nodes were found to express CD271. It has been reported that NGFR interaction with its ligand, NGF, may play a role in immunoregulation. NGF may function as a B-cell growth factor.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD271
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon C40-1457
Conjugado PE
Descripción NGFR; NTR; p75NTR; p75 ICD; TNFRSF16; TNR16; LNGFR
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.