Learn More
Invitrogen™ CD268 (BAFF Receptor) Polyclonal Antibody

Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse BAFF R and less than 2% cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, rhRANK, rhTNF RI, and rhTNF RII is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Spécification
Spécification
| Antígeno | CD268 (BAFF Receptor) |
| Aplicaciones | Flow Cytometry, Immunohistochemistry (Paraffin), Neutralization, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 5% trehalose and No Preservative |
| génica | TNFRSF13C |
| N.º de referencia del gen | Q96RJ3 |
| Alias de gen | 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C |
| Símbolos de los genes | TNFRSF13C |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.