Learn More
Invitrogen™ CD267 (TACI) Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Marca:  Invitrogen™ PA547192
Código de producto. 15567682
  • 510.00€ / 100 microgramos
Descripción
In direct ELISAs, less than 5% cross-reactivity with recombinant mouse TACI and recombinant human BCMA is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CD267 (TACI)
Polyclonal
Unconjugated
TNFRSF13B
1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Goat
Antigen affinity chromatography
RUO
23495
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunohistochemistry (Paraffin), Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
O14836
TNFRSF13B
Mouse myeloma cell line NS0-derived recombinant human TACI/TNFRSF13B Ser2-Thr166.
100 μg
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.