Learn More
Invitrogen™ CD267 (TACI) Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Marca:  Invitrogen™ PA547192
Código de producto. 15567682
  • 510.00€ / 100 microgramos
Descripción
In direct ELISAs, less than 5% cross-reactivity with recombinant mouse TACI and recombinant human BCMA is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CD267 (TACI)
Polyclonal
Unconjugated
TNFRSF13B
1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Goat
Antigen affinity chromatography
RUO
23495
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunohistochemistry (Paraffin), Neutralization, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
O14836
TNFRSF13B
Mouse myeloma cell line NS0-derived recombinant human TACI/TNFRSF13B Ser2-Thr166.