missing translation for 'onlineSavingsMsg'
Learn More

CD263 (TRAIL-R3) Polyclonal Antibody, Invitrogen™

Código de producto. 11518661
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11518661 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. View return policy
Product Code. 11518661 Supplier Invitrogen Supplier No. PA1956

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-956 detects decoy receptor 1 (DcR1) from human and rat tissues and cells. PA1-956 has been successfully used in FACS and Western blot procedures. By Western blot, this antibody detects an ∽30 kDa protein representing DcR1 from HEK293 cell lysate. PA1-956 immunizing peptide corresponds to amino acid residues 24-35 of human DcR1. PA1-956 immunizing peptide (Cat. No. PEP-047) is available for use in neutralization and control experiments.

DcR1 / TRAIL-R3 / TRID / LIT is one of the two putative decoy receptors identified for TRAIL, one of the members of TNF family of apoptosis inducing proteins. The other putative decoy receptor is called DcR2 / TRUNDD / TRAIL-R4. DcR1 is attached to the cell surface through glycophospholipid anchor. It has the extracellular TRAIL binding domain but lacks the cytoplasmic domain to induce apoptotic signal. Hence overexpression of DcR1 inhibits the TRAIL induced apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD263 (TRAIL-R3)
Aplicaciones Flow Cytometry, Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
génica TNFSF10
N.º de referencia del gen O14798, P50591
Alias de gen CD263, DCR1, LIT, MGC149501, MGC149502, TRAILR3, TRID
Símbolos de los genes TNFRSF10C, TNFSF10
Especie del huésped Rabbit
Inmunógeno Synthetic Peptide: Y(24) S A T T A R Q E E V P(35) C
Método de purificación Antigen affinity chromatography
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 246775, 8743, 8794
Especies diana Human, Rat
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.