missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD26 Monoclonal Antibody (202.36)
GREENER_CHOICE

Product Code. 11540722
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microgramos
Product Code. Cantidad unitSize
11540722 100 μg 100 microgramos
1 options
This item is not returnable. View return policy

Product Code. 11540722

Brand: Invitrogen™ MA135147

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

202-36 Reacts with human CD26 (Dipeptidyl peptidase IV), a 110 kDa protein which occurs in the cell membrane of T-lymphocyte and is upregulated after activation. CD26 is also present on endothelial cells and also expressed on activated B-cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Crosslinking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production. In Western blotting, this mAb reacts with only glycosylated CD26, but not with deglycosylated froms. It does not prevent ADA binding to CD26. The mAb was clustered at the VIth WLDA.

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of malignant transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD26
Aplicaciones Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Clasificación Monoclonal
Clon 202.36
Concentración 0.1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
génica DPP4
N.º de referencia del gen P14740, P27487
Alias de gen ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10
Símbolos de los genes DPP4
Especie del huésped Mouse
Inmunógeno T-cell clone.
Método de purificación Purified
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 1803, 25253
Especies diana Human, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.