missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD25 Monoclonal Antibody (YTH906.9HL)
GREENER_CHOICE

Código de producto. 15343961
Change view
Click to view available options
Cantidad:
200 μg
Tamaño de la unidad:
200 microgramos
Código de producto. Cantidad unitSize
15343961 200 μg 200 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15343961

Marca: Invitrogen™ MA517677

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

MA5-17677 targets CD25/IL-2R alpha chain in FACS and WB applications and shows reactivity with Human samples. The MA5-17677 immunogen is rat monoclonal antibody raised against activated human T lymphocytes.

CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD25
Aplicaciones Flow Cytometry, Western Blot
Clasificación Monoclonal
Clon YTH906.9HL
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.02% sodium azide
génica IL2RA
N.º de referencia del gen P01589
Alias de gen ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR
Símbolos de los genes IL2RA
Especie del huésped Rat
Inmunógeno Rat monoclonal antibody raised against activated human T lymphocytes.
Método de purificación Protein G
Cantidad 200 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3559
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.