missing translation for 'onlineSavingsMsg'
Learn More

CD23 Mouse anti-Human, Brilliant Violet 650, Clone: EBVCS-5 (also known as EBVCS 5), BD Optibuild™

Código de producto. 15838259
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15838259 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15838259 Proveedor BD Biosciences N.º de proveedor 743429

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The EBVCS-5 antibody specifically binds to human CD23, the low affinity receptor for human IgE (Fc epsilon RII orFcεRII), which is also known as C-type lectin domain family 4 member J (CLEC4J). CD23 is expressed as a ñ45 kDa type II membrane glycoprotein that is present at low density on most normal B lymphocytes and at higher levels on activated B lymphocytes, Epstein-Barr virus (EBV)-transformed lymphoblasts, chronic lymphocytic leukemia (CLL) cells of B-lymphocyte origin, and tonsillar B lymphocytes. CD23 expression is induced on B cells by interleukin-4 (IL-4) and is lost after isotype switching to IgA, IgG, or IgE. The CD23 antigen is not present on immature bone marrow B lymphocytes or on T lymphocytes. CD23 may also be differentially expressed on monocytes, macrophages, eosinophils, platelets, dendritic cells, and Langerhans cells. CD23 can mediate IgE-dependent cytotoxicity and phagocytosis by macrophages and eosinophils. Soluble CD23 (sCD23) can be released by CD23-positive cells as a result of proteolytic cleavage of membrane CD23. Larger fragments of sCD23 (eg, 25-37 kDa) retain their IgE-binding capacity whereas smaller fragments (ie, ≤ 12 kDa) do not. Soluble CD23 may have immunoregulatory effects on the growth and differentiation of B cells and other cell types.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD23
Clasificación Monoclonal
Clon EBVCS-5 (also known as EBVCS 5)
Concentración 0.2mg/mL
Conjugado Brilliant Violet 650
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen CD23A, Fc-epsilon-RII, FCER2, FCE2, IGEBF, BLAST-2, CLEC4J, Leu-20
Especie del huésped Mouse
Inmunógeno Epstein-Barr Virus Transformed Cell Line
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.