Learn More
CD221 Mouse anti-Human, PE, Clone: 1H7, BD
Descripción
The 1H7 monoclonal antibody specifically binds to the α subunit of the human insulin-like growth factor-I (IGF-I) receptor that is also known as CD221. CD221 is a receptor tyrosine kinase that has high affinity for IGF-1 and low affinity for insulin and IGF-2. CD221 is synthesized as a 180 kDa precursor molecule that is processed into two subunits, a 135 kDa α subunit which is disulfide-linked to a 90 kDa β subunit. The mature IGF-IR complex is comprised of two extracellular α subunits and two transmembrane β subunits that each possesses a tyrosine kinase domain. When bound by ligand, the IGF-1 receptor autophosphorylates multiple tyrosine sites in its β chains and phosphorylates other signaling molecules that regulate cell growth, development, and neoplastic transformation. CD221 is structurally and functionally similar to the insulin receptor (aka, CD220) and is expressed on a variety of human hematopoietic and non-hematopoietic cells. The 1H7 antibody does not crossreact with the human insulin receptor and can block IGF-I- or IGF-II-stimulated cell growth.
Especificaciones
Especificaciones
| Antígeno | CD221 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 1H7 |
| Conjugado | PE |
| Descripción | IGF1R; IGF-I receptor; IGFIR; IGFR; Insulin-like growth factor 1 receptor |
| Formulación | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Especie del huésped | Mouse |
| Inmunógeno | Purified Human Placental IGF-I Receptor |
| Método de purificación | Affinity Purified |
| Mostrar más |
For Research Use Only.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.