missing translation for 'onlineSavingsMsg'
Learn More

CD212 Mouse anti-Human, Brilliant Violet 421, Clone: 2.4E6 (also known as 2-4E6), BD Optibuild™

Código de producto. 15828849
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15828849 50 μg cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15828849

Marca: BD Biosciences 744202

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The 2.4E6 monoclonal antibody specifically binds to CD212, which is also known as the Interleukin-12 Receptor beta-1 chain (IL12RB1/IL-12RB1/IL-12Rβ1). CD212 serves as a subunit for the functional IL-12 Receptor (IL-12Rβ1-IL-12Rβ2) and IL-23 Receptor (IL-12Rβ1-IL-23R) complexes. CD212 is expressed on T cells, B cells, or NK cells. CD212 is a type I transmembrane glycoprotein that belongs to the hemopoietin receptor superfamily. CD212 is responsible for binding to the IL-12 p40 subunit that is common to the heterodimeric IL-12 and IL-23 cytokines. IL-12 and IL-23 are immunomodulatory cytokines produced mainly by antigen-presenting cells (APC) including monocytes and dendritic cells. These cytokines have overlapping as well as distinct biological activities.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD212
Clasificación Monoclonal
Clon 2.4E6 (also known as 2-4E6)
Concentración 0.2mg/mL
Conjugado Brilliant Violet 421
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen IL-12R beta 1, IL-12R-BETA1, IL-12 receptor β1, IMD30
Especie del huésped Mouse
Inmunógeno PHA-activated Human PBMC
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.