missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD21 Monoclonal Antibody (CC51), PE

Código de producto. 16342594
Change view
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
paquete de 100
Código de producto. Cantidad unitSize
16342594 100 Tests paquete de 100
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16342594

Marca: Invitrogen™ MA528322

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Mouse anti Bovine CD21 monoclonal antibody, clone CC51 recognizes the bovine homologue of the human CD21 cell surface antigen, a 145 kDa single pass type I membrane glycoprotein containing multiple sushi domains. Reconstitute with 1.0 mL distilled water.

CD21 (complement receptor 2, CR2, C3D receptor, EBV receptor) binds C3 complement fragments, especially its breakdown fragments, which remain covalently attached to complement activating surfaces or antigen. CD21 has important roles in uptake and retention of immunocomplexes, survival of memory B cells and in development and maintenance of the humoral response to T-dependent antigens. CD21 also serves as a key receptor for Epstein-Barr virus binding and is involved in targeting prions to follicular dendritic cells and expediting neuroinvasion following peripheral exposure to prions. A soluble form of the CD21 (sCD21) is shed from the lymphocyte surface and retains its ability to bind respective ligands. CD21 functions as receptor for C3d, C3dg and iC3b complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. Genetic variations in the CD21 gene are associated with susceptibility to systemic lupus erythematosus type 9 (SLEB9). Alternatively, spliced transcript variants encoding different isoforms of CD21 have been found.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD21
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon CC51
Concentración 1 mg/mL
Conjugado PE
Formulación PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
génica CR2
N.º de referencia del gen 0
Alias de gen C3b/C4b receptor; C3-binding protein; C3BR; C3DR; C4BR; CD21; CD35; CD35 antigen; cell surface receptor for C3d; complement C3b/C4b receptor 1 (Knops blood group); complement C3d receptor; Complement C3d receptor (C3DR); complement C3d receptor 2; complement component (3b/4b) receptor 1 (Knops blood group); complement component (3d/Epstein Barr virus) receptor 2; complement component 3b/4b receptor 1 (Knops blood group); complement component 3d receptor 2; complement component receptor 2; complement receptor 1 long isoform; complement receptor 2; complement receptor type 1; complement receptor type 1-like protein; complement receptor type 2; Complement Receptor type 2 (CR2); CR; CR1; Cr-1; CR1-L; Cr2; Cr-2; CVID7; EBV receptor; EBV-R; Epstein-Barr virus receptor; EVBR; expressed in B lymphocytes; KN; Knops blood group antigen; LOW QUALITY PROTEIN: complement receptor type 1; SLEB9
Símbolos de los genes CR2
Especie del huésped Mouse
Inmunógeno Bovine (Friesian cattle) mesenteric lymph node cells.
Método de purificación Protein A
Cantidad 100 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 407126
Especies diana Bovine
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2b
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.