Learn More
CD206 Mouse anti-Human, FITC, Clone: 15-2, Proteintech
Código de producto. 17254866 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 Tests
Tamaño de la unidad:
100 reacciones
17254866 100 Tests 100 reacciones
1 options
Código de producto. 17254866
Marca: Proteintech FITC65155100TESTS
Mouse Monoclonal Antibody
CD206, also named as MMR, CLEC13D and MRC1, is a type I membrane receptor that mediates the endocytosis of glycoproteins by macrophages. CD206 has been shown to bind high-mannose structures on the surface of potentially pathogenic viruses, bacteria, and fungi so that they can be neutralized by phagocytic engulfment. CD206 is a 170 kDa transmembrane protein which contains 5 domains: an amino-terminal cysteine-rich region, a fibronectin type II repeat, a series of eight tandem lectin-like carbohydrate recognition domains (responsible for the recognition of mannose and fucose), a transmembrane domain, and an intracellular carboxy-terminal tail. It is expressed on most tissue macrophages, in vitro derived dendritic cells, lymphatic and sinusoidal endothelia.
Especificaciones
Antígeno CD206
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 15-2
Concentración 4 μL/Test
Conjugado FITC
Especie del huésped Mouse
Cantidad 100 Tests
Primario o secundario Primary
ID de gen (Entrez) 4360
Especies diana Human, Mouse
Formulario Liquid
Isotype IgG1 κ