missing translation for 'onlineSavingsMsg'
Learn More

CD205 Monoclonal Antibody (205yekta), NovaFluor™ Red 685, eBioscience™, Invitrogen™

Código de producto. 17852964
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17852964 25 μg 25 microgramos
17842874 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17852964 Proveedor Invitrogen N.º de proveedor M051T02R02

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The 205yekta antibody reacts with mouse CD205, also known as DEC-205 (dendritic and epithelial cells, 205 kDa). CD205 is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 has been shown to bind to PE-Cy5.5 antibody and streptavidin conjugates; therefore, this format should not be used on CD205+ dendritic cells. Applications Tested: This 205yekta antibody has been tested by flow cytometric analyis of mouse bone marrow cells. This can be used at less than or equal to 0.2 ug per test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100 uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Use 5uL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 637 nm; Emission: 685 nm; Laser: 633-640 nm (Red) Laser

CD205 (LY25, Lymphocyte Antigen 75) is an integral membrane glycoprotein involved in antigen uptake, trafficking and presentation that improves the induction of antigen-specific T cell immunity. CD205 is highly expressed by CD8+ dendritic cells (DCs) and also expressed at different levels by bone marrow Gr1+ cells, Langerhans cells, (BMDC) bone marrow derived DCs and thymic epithelial cells. CD205 acts as an endocytic receptor that directs captured antigens to the extracellular space towards a specialized antigen processing region. Further, CD205 also prompts the reduced proliferation of B lymphocytes. Diseases associated with CD205 dysfunction include pneumonic plague and lipid pneumonia.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD205
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 205yekta
Concentración 0.1 mg/mL
Conjugado NovaFluor Red 685
Formulación PBS with 0.09% sodium azide; pH 7.2
génica LY75
N.º de referencia del gen Q60767
Alias de gen CA-IX; CD205; CLEC13B; C-type lectin domain family 13 member B; DCL-1 fusion; DEC205; DEC-205; DEC-205/CD205 protein; gp200; gp200-MR6; group VI C-type animal lectin; LY75; ly-75; Lymphocyte antigen 75; membrane antigen MN; P54/58N; pMW1; RGD1565962; RP23-10L17.4
Símbolos de los genes LY75
Especie del huésped Rat
Método de purificación Affinity chromatography
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17076
Especies diana Mouse
Contenido y almacenamiento 4° C, store in dark, DO NOT FREEZE!
Formulario Liquid
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.