missing translation for 'onlineSavingsMsg'
Learn More

CD205 (DEC-205) Rat anti-Mouse, Brilliant Violet 421, Clone: V18-949, BD

Código de producto. p-7082876
Change view
Click to view available options
Cantidad:
25 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15824999 25 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15824999 Proveedor BD Biosciences N.º de proveedor 566376

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The V18-949 monoclonal antibody specifically recognizes CD205 which is also known as Dendritic and thymic Epithelial Cells, 205 kDa (DEC-205). CD205 (DEC-205) is a type 1 transmembrane glycoprotein that is encoded by Ly75 (lymphocyte antigen 75). CD205 (DEC-205) is highly expressed by both CD8+ and CD103+ dendritic cell subsets and Langerhans cells. It is variably expressed by some other dendritic cell types, Gr1+ (Ly-6G and Ly-6C) bone marrow cells, B cells, macrophages, granulocytes, and mucosal and thymic cortical epithelial cells. CD205 (DEC-205) belongs to the C-type lectin receptor family and functions as a pattern recognition receptor and an endocytic receptor that participates in the cellular processing and presentation of foreign or self antigens to initiate immunity or tolerance.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD205 (DEC-205)
Clasificación Monoclonal
Clon V18-949
Concentración 0.2mg/mL
Conjugado Brilliant Violet 421
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Alias de gen Cd205, DEC-205, DEC205, Ly-75, Ly75, Lymphocyte antigen 75
Especie del huésped Rat
Inmunógeno Mouse CD205 (DEC-205)
Método de purificación Affinity Purified
Cantidad 25 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Mouse
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.