missing translation for 'onlineSavingsMsg'
Learn More

CD201 Rat anti-Human, Brilliant Violet 711, Clone: RCR-252, BD Optibuild™

Código de producto. 15808349
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
15808349 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15808349 Proveedor BD Biosciences N.º de proveedor 743555

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

The RCR-252 monoclonal antibody specifically binds to the endothelial cell protein C receptor (EPCR), which is also known as CD201. CD201 is a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. CD201 binds activated protein C, a serine protease that is involved in regulating blood coagulation. EPCR is reportedly expressed on several malignant cell lines. Malignant cell lines that express EPCR can show significant levels of protein C activation and this activation can be inhibited by anti-EPCR antiboy.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD201
Clasificación Monoclonal
Clon RCR-252
Concentración 0.2mg/mL
Conjugado Brilliant Violet 711
Formulación Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gen EPCR, Activated protein C receptor, APC receptor, PROCR, centrocyclin
Especie del huésped Rat
Inmunógeno EPCR-positive RE-1 Cells
Método de purificación Affinity Purified
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.