missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Antibody (divozilimab) [Janelia Fluor™ 635], Novus Biologicals™
Description
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antígeno | CD20 |
| Aplicaciones | Flow Cytometry, ELISA, Functional Assay |
| Clasificación | Monoclonal |
| Clon | divozilimab |
| Conjugado | Janelia Fluor 635 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 |
| Especie del huésped | Human |
| Inmunógeno | CD20 |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?