missing translation for 'onlineSavingsMsg'
Learn More

CD2, Mouse anti-Human, Polyclonal Antibody, Abnova™

Código de producto. 16135784
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
Código de producto. Cantidad unitSize
16135784 50 μg 50 microgramos
1 options
This item is not returnable. View return policy

Product Code. 16135784

Brand: Abnova H00000914B02P.50ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MSFPCKFVASFLLIFNVSSKGAVSKEITNALETWGALGQDINLDIPSFQMSDDIDDIKWEKTSDKKKIAQFRKEKETFKEKDTYKLFKNGTLKIKHLKTDDQDIYKVSIYDTKGKNVLEKIFDLKIQERVSKPKISWTCINTTLTCEVMNGTDPELNLYQDGKHLKLSQRVITHKWTTSLSAKFKCTAGNKVSKESSVEPVSCPEKGLDIYLIIGICGGGSLLMVFVALLVFYITKRKKQRSRRNDEELETRAHRVATEERGRKPQQIPASTPQNPATSQHPPPPPGHRSQAPSHRPPPPGHRVQHQPQKRPPAPSGTQVHQQKGPPLPRPRVQPKPPHGAAENSLSPSSN

Specifications

Antígeno CD2
Aplicaciones Immunohistochemistry (Paraffin), Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Formulación In 1x PBS, pH 7.4
génica CD2 molecule
N.º de referencia del gen BC033583
Alias de gen SRBC/T11
Símbolos de los genes CD2
Especie del huésped Mouse
Inmunógeno CD2 (AAH33583, 1 a.a. ∼ 351 a.a) full-length human protein.
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 914
Especies diana Human
Contenido y almacenamiento Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Tipo de producto Antibody
Formulario Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.