missing translation for 'onlineSavingsMsg'
Learn More

CD2 Mouse anti-Human, BV421, Clone: L303.1 (also known as L303), BD OptiBuild™

Product Code. 15819329
Change view
Click to view available options
Cantidad:
50 μg
Unit Size:
cada uno
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
15819329 50 μg cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819329 Supplier BD Biosciences Supplier No. 744826

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD2
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon L303.1 (also known as L303)
Concentración 0.2 mg/mL
Conjugado Brilliant Violet 421
Especie del huésped Mouse
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Refrigerate (2 to 8°C)
Formulario Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.