missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1d Antibody (1B1) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP1-43461JF525
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD1d Monoclonal antibody specifically detects CD1d in Hamster,Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation
Especificaciones
| CD1d | |
| Monoclonal | |
| Janelia Fluor 525 | |
| antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D | |
| The immunogen for this antibody was CD1d. | |
| 0.1 mL | |
| Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience | |
| 912 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| 1B1 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Hamster, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction