missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD1c Monoclonal Antibody (L161), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™

Código de producto. 30055252
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
paquete de 100
paquete de 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30055252 25 Tests paquete de 25
30055057 100 Tests paquete de 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30055252 Proveedor Invitrogen™ N.º de proveedor H046T02B08A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

CD1c is a member of the CD1 family of transmembrane glycoproteins, which are structurally related to major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. This family of proteins is involved in the presentation of lipid and glycolipid antigens, both of self and microbial origin, to T cells during the adaptive immune response. CD1c is expressed on some circulating and marginal zone B cells, as well as in lymph nodes and germinal centers. It plays a crucial role in presenting lipid antigens, such as microbial fatty acids, to effector T cells. The protein encoded by the CD1c gene localizes to late endosomes and lysosomes, utilizing a tyrosine-based motif in its cytoplasmic tail for targeting. Vesicular acidification is required for CD1c to bind lipid antigens effectively. The human genome contains five CD1 family genes organized in a cluster on chromosome 1, with each member differing in cellular localization and specificity for particular lipid ligands. CD1c undergoes alternative splicing, resulting in three different isoforms: soluble, membrane-bound, and cytoplasmic/soluble isoforms, highlighting its functional diversity in immune processes.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno CD1c
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon L161
Concentración 0.2 μg/Test
Conjugado NovaFluor Blue 660-120S
Formulación PBS with 0.09% sodium azide; pH 7.2
génica CD1C
N.º de referencia del gen P29017
Alias de gen BDCA1; canCD1c; CD1; CD1A; CD1C; CD1C antigen, c polypeptide; CD1c molecule; cortical thymocyte antigen CD1C; differentiation antigen CD1-alpha-3; R7; RP11-101J8.3; T-cell surface glycoprotein CD1c
Símbolos de los genes CD1C
Especie del huésped Mouse
Método de purificación Affinity chromatography
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 911
Especies diana Human
Contenido y almacenamiento 4°C, store in dark, DO NOT FREEZE!
Tipo de producto Antibody
Formulario Liquid
Formato Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.