missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD199 (CCR9) Monoclonal Antibody (9B1), PE

Descripción
Rat anti Mouse CDw199 antibody, clone 9B1 detects mouse chemokine receptor 9 (CCR9), also known as CDw199. Reconstitute by adding 1 mL distilled water.
The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.
Especificaciones
Especificaciones
| Antígeno | CD199 (CCR9) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 9B1 |
| Concentración | Conc. Not Determined |
| Conjugado | PE |
| Formulación | PBS with 1% BSA, 5% sucrose and 0.09% sodium azide |
| génica | Ccr9 |
| N.º de referencia del gen | Q9WUT7 |
| Alias de gen | A130091K22Rik; C-C chemokine receptor type 9; C-C CKR-9; C-C motif chemokine receptor 9; CC-CKR-9; Ccr9; CCR-9; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; chemokine C-C receptor 10; Cmkbr10; G protein-coupled receptor 28; GPR28; GPR-9-6; G-protein coupled receptor 28 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?