missing translation for 'onlineSavingsMsg'
Learn More

CD197, CCR7 Rat anti-Human, APC-R700, Clone: 3D12, BD Horizon™

Código de producto. p-7068068
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15864569 25 Tests cada uno
15854569 100 Tests cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15864569

Marca: BD Biosciences 565868

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rat Monoclonal Antibody

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD197, CCR7
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 3D12
Conjugado APC-R700
Descripción Vol. Per test: 5μL
Formulación Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Alias de gen CCR7, BLR-2, EBI-1, CMKBR7
Especie del huésped Rat
Inmunógeno Human CCR7 protein
Cantidad 25 Tests
Estado normativo RUO
Disciplina de investigación Cell Biology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
Formulario Purified
Isotype IgG2a κ
Mostrar más Mostrar menos

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.