missing translation for 'onlineSavingsMsg'
Learn More

CD183 Mouse anti-Human, PE, Clone: 1C6/CXCR3, BD

Código de producto. 15878848
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15878848 100 Tests cada uno
15820609 25 Tests cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15878848

Marca: BD Biosciences 557185

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

The 1C6/CXCR3 monoclonal antibody specifically binds to human CD183, also known as the CXCR3 chemokine receptor. CD183 is a 40-41 kDa seven-transmembrane protein and member of the G protein-coupled receptor family. CD183 is expressed primarily on activated T cells that infiltrate inflammatory sites. It has also been detected on some circulating T cells, B cells, and NK cells. Reports show that some CXCR3-positive T cells also express CCR5 and are mostly CD45RO-positive cells. Three ligands for CXCR3 have been identified. They are CXCL9 (Mig/monokine induced by interferon-γ), CXCL10 (IP-10/interferon-γinducible 10-kD protein), and CXCL11 (I-TAC/interferon-inducible T-cell alpha chemoattractant). These chemokines are produced by a variety of cells upon stimulation by IFN-γand interact with CXCR3 to mediate T-cell chemotaxis. This reagent has been reported to be suitable for immunohistochemical staining of acetone-fixed, frozen sections and/or formalin-fixed, paraffin-embedded tissue sections with citrate pretreatment. Clone 1C6/CXCR3 also cross reacts with a subset of peripheral blood lymphocytes of baboon, and both rhesus and cynomolgus macaque monkeys. The distribution of lymphocytes is similar to that observed with CD183-positive peripheral blood lymphocytes from normal human donors. CXCR3 has been clustered as CD183 in the VIIth HLDA workshop.

TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno CD183
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 1C6/CXCR3
Conjugado PE
Descripción CXCR3; C-X-C chemokine receptor type 3; GPR9; IP10R; MigR; CKRL2; CMKAR3
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human CXCR3 Peptide
Método de purificación Affinity Purified
Cantidad 100 Tests
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Mostrar más Mostrar menos

For Research Use Only.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.