Learn More
Invitrogen™ CD18 Chimeric Recombinant Rabbit Monoclonal Antibody (1B4)

Descripción
Specificity: Specific to the beta chain of the leukocyte integrin beta-2 family. This is found in LFA-1, Mac-1 and p150.95 among others. Binds an epitope distinct from the C3bi binding site. Has been shown to recognise both the human and Rhesus monkey antigen. Preparation: Blood was obtained from 4 helathy donors and the mononuclear cells harvested and grown. A BALB/c mouse was injected with 10^6 monocytes three times at monthly intervals, and four times with 5x10^6 monocytes oon each of the four days before fusion. The spleen cells were then fused to P3U1 myeloma cells to generate hybridomas and RIA was used to identify lines of interest.
Especificaciones
Especificaciones
| Antígeno | CD18 Chimeric |
| Aplicaciones | ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 1B4 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.02% ProClin 300 |
| génica | ITGB2 |
| N.º de referencia del gen | P05107 |
| Alias de gen | 2E6; AI528527; antigen CD18; B-2 integrin; CD18; CD18 leukocyte adhesion molecule; CD18 subunit; cell surface adhesion glycoprotein (LFA-1/CR3/P150,959 beta subunit precursor); cell surface adhesion glycoproteins LFA-1/CR3/p150,95 subunit beta; complement component 3 receptor 3 and 4 subunit; complement receptor C3 beta-subunit; complement receptor C3 subunit beta; integrin beta 2; integrin beta chain, beta 2; integrin beta-2; integrin subunit beta 2; integrin, beta 2; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1; macrophage antigen 1 (mac-1) beta subunit); integrin, beta 2 (antigen CD18 subunit (p95), lymphocyte function-associated antigen 1, integrin B2); integrin, beta 2 (complement component 3 receptor 3 and 4 subunit); ITGB2; LAD; LCAMB; Leu-CAM receptor; leukocyte cell adhesion molecule CD18; leukocyte surface protein; leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C; Lfa1; LFA-1; lymphocyte function associated antigen 1; MAC-1; Mac-1 beta; macrophage antigen 1 (mac-1) beta subunit); macrophage antigen-1 beta; MF17; MFI7 |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.