Learn More
Invitrogen™ CD179a Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Marca:  Invitrogen™ PA542149
Código de producto. 15565282
  • 530.00€ / 100 microlitros
Descripción
Peptide sequence: TIRLTCTLRN DHDIGVYSVY WYQQRPGHPP RFLLRYFSQS DKSQGPQVPP Sequence homology: Cow: 92%; Dog: 86%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 79%.
The protein encoded by this gene belongs to the immunoglobulin superfamily and is expressed selectively at the early stages of B cell development, namely, in proB and early preB cells. This gene encodes the iota polypeptide chain that is associated with the Ig-mu chain to form a molecular complex which is expressed on the surface of pre-B cells. The complex is thought to regulate Ig gene rearrangements in the early steps of B-cell differentiation.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
CD179a
Polyclonal
Unconjugated
VPREB1
CD179 antigen-like family member A; CD179a; IGI; IGLL1; IGVPB; Immunoglobulin iota chain; Immunoglobulin lambda Vpreb1 chain; immunoglobulin lambda-1 light chain; immunoglobulin lambda-like polypeptide 5; LOC100735142; pre-B lymphocyte 1; pre-B lymphocyte gene 1; Protein VPreB1; surrogate light chain; v(pre)B protein; VPREB; VpreB protein; Vpreb1; Vpreb-1; V-set pre-B cell surrogate light chain 1
Rabbit
Affinity Chromatography
RUO
7441
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P12018
VPREB1
Synthetic peptide directed towards the middle region of human VPREB1.
100 μL
Primary
Human
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones