missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD16b (FCGR3B) Monoclonal Antibody (C1), Invitrogen™
Descripción
CD16 (FCGR3A) is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by NK cells and some T cells, and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC by NK cells. Human CD16 is expressed in two forms - FCGR3A and FCGR3B. FCGR3A is associated with the FcepsilonRI-gamma subunit and is responsible for antibody-dependent NK cell cytotoxicity. Mast cell FCGR3A is associated, with FcepsilonRI-beta subunit. Besides IgG, FCGR3A can be triggered also by oligomeric IgE. FCGR3B is a GPI-linked monomeric receptor expressed on neutrophils and is involved in their activation and induction of a pro-adhesive phenotype. Diseases associated with CD16 dysfunction include immunodeficiency 20 and systemic lupus erythematosus.
Especificaciones
Especificaciones
| Antígeno | CD16b (FCGR3B) |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | C1 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| génica | FCGR3B |
| N.º de referencia del gen | O75015 |
| Alias de gen | CD16; CD16B; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor IIIb; FCG3; Fc-gamma receptor IIIb (CD 16); Fc-gamma RIII; fc-gamma RIIIb; Fc-gamma RIII-beta; FCGR3; FCGR3B; FcR-10; FCRIII; FCRIIIb; IGFR3; IgG Fc receptor III-1; low affinity immunoglobulin gamma Fc region receptor III-B |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?