missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD169 (Siglec-1) Monoclonal Antibody (3B11/11), FITC

Descripción
Mouse anti Pig CD169, clone 3B11/11 Is specific for porcine CD169, also known as sialoadhesin or Siglec-1, a member of the sialic acid binding immunoglobulin-like lectin (Siglec) family.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.
Especificaciones
Especificaciones
| Antígeno | CD169 (Siglec-1) |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 3B11/11 |
| Concentración | 0.1 mg/mL |
| Conjugado | FITC |
| Formulación | PBS with 1% BSA and 0.09% sodium azide |
| génica | SIGLEC1 |
| N.º de referencia del gen | A7LCJ3 |
| Alias de gen | Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?