Learn More
Invitrogen™ CD163L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5116626
Descripción
Positive Samples: Rat heart, Rat lung Immunogen sequence: DCLHQNDVSV ICSDGADLEL RLADGSNNCS GRVEVRIHEQ WWTICDQNWK NEQALVVCKQ LGCPFSVFGS RRAKPSNEAR DIWINSISCT GNESALWDCT YDGKAKRTCF RRSDAGVICS DKADLDLRLV GAHSPCYGRL EVKYQGEWGT VCHDRWSTRN AAVVCKQLGC GKPMHVFGMT YFKEASGPIW LDDVSCIGNE S.
This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily. Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system. The SRCR family is defined by a 100-110 amino acid SRCR domain, which may mediate protein-protein interaction and ligand binding. The encoded protein contains twelve SRCR domains, a transmembrane region and a cytoplasmic domain. Alternatively spliced transcript variants encoding different isoforms have been described but their full-length nature has not been determined.
Especificaciones
| CD163L1 | |
| Polyclonal | |
| Unconjugated | |
| CD163L1 | |
| CD163 antigen B; CD163 antigen-like 1; CD163 molecule like 1; CD163 molecule-like 1; CD163B; CD163L1; M160; SCARI2; scavenger receptor cysteine-rich type 1 protein M160; UNQ6434/PRO23202 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-550 of human CD163L1 (NP_7776012). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9NR16 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283316 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.