missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD162 Mouse, Clone: PSGL1/1601, enQuireBio™
Specifications
Specifications
| Antígeno | CD162 |
| Aplicaciones | Immunohistochemistry |
| Localización celular | Cell Surface |
| Clasificación | Monoclonal |
| Clon | PSGL1/1601 |
| Concentración | 200μg/mL |
| Conjugado | Unconjugated |
| Formulación | 10mM PBS with 0.05% BSA and 0.05% Azide |
| N.º de referencia del gen | NP_001193538 |
| Símbolos de los genes | SELPLG |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?