missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD160 Recombinant Rabbit Monoclonal Antibody (275)

Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen™ MA536025
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD160 Recombinant Monoclonal Antibody for Flow
CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.
Especificaciones
| CD160 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O95971 | |
| CD160 | |
| Recombinant Human CD160 Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 275 | |
| Unconjugated | |
| CD160 | |
| AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido