missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, Alexa Fluor 647, Clone: 3G8, BD

Código de producto. 15891269
Change view
Click to view available options
Cantidad:
100 Tests
25 Tests
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
15891269 25 Tests cada uno
15819008 100 Tests cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Artikelnummer. 15891269

Brand: BD Biosciences 561724

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

The 3G8 monoclonal antibody specifically binds to the 50-65 kDa transmembrane form of the IgG Fc Receptor (Fc γRIII), a human NK cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. The 3G8 antibody blocks the binding of soluble immune complexes to granulocytes. The 3G8 antibody is reported (Vossebeld et al ., 1997) to increase intracellular calcium levels in human neutrophils by interacting with both Fc γRIIa and Fc γRIIIb molecules. This antibody has also been reported to induce homotypic neutrophil aggregation.

This clone also cross-reacts with a subset of peripheral blood lymphocytes and monocytes, but not granulocytes, of baboon and both rhesus and cynomologus macaque monkeys. Multi-color analysis reveals that the distribution on lymphocytes is similar to that found in human studies with the majority of CD16-positive lymphocytes being both CD3 and CD20 negative.

TRUSTED_SUSTAINABILITY

Tekniske data

Antígeno CD16
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 3G8
Conjugado Alexa Fluor 647
Descripción FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Formulación Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Especie del huésped Mouse
Inmunógeno Human polymorphonuclear leukocytes
Método de purificación Affinity Purified
Cantidad 25 Tests
Estado normativo RUO
Primario o secundario Primary
Especies diana Baboon, Cynomolgus, Human, Rhesus Monkey
Contenido y almacenamiento Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.