missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD147 Recombinant Rabbit Monoclonal Antibody (23GB1400), Invitrogen™

Descripción
CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and malignant tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
Especificaciones
Especificaciones
| Antígeno | CD147 |
| Aplicaciones | Flow Cytometry, Western Blot, Immunocytochemistry |
| Clasificación | Recombinant Monoclonal |
| Clon | 23GB1400 |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| génica | BSG |
| N.º de referencia del gen | P35613 |
| Alias de gen | 5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?